Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Ohaus
®
PR Analytical Balance
Products
Product Category
balances (6)
Manufacturer
Weighing Capacity (g)
Package Size
Filter Products
Showing 1-6 of 6
Compare
Product Number
Description
Pricing
OH30430070
Ohaus
®
PR Analytical Balance,
Model PR224/E, weighing capacity 220 g, resolution: 0.1 mg, AC/DC input 230 V, universal plug set
Expand
View Pricing
OH30430068
Ohaus
®
PR Analytical Balance,
Model PR124/E, weighing capacity 120 g, resolution: 0.1 mg, AC/DC input 230 V, universal plug set
Expand
View Pricing
OH30430069
Ohaus
®
PR Analytical Balance,
Model PR224, weighing capacity 220 g, resolution: 0.1 mg, AC/DC input 230 V, universal plug set
Expand
View Pricing
OH30430067
Ohaus
®
PR Analytical Balance,
Model PR124, weighing capacity 120 g, resolution: 0.1 mg, AC/DC input 230 V, universal plug set
Expand
View Pricing
OH30524385
Ohaus
®
PR Analytical Balance,
Model PR224M, weighing capacity 220 g, resolution: 0.1 mg, AC/DC input 230 V, universal plug set
Expand
View Pricing
OH30524384
Ohaus
®
PR Analytical Balance,
Model PR124M, weighing capacity 120 g, resolution: 0.1 mg, AC/DC input 230 V, universal plug set
Expand
View Pricing