Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Ohaus
®
Explorer
®
Semi-Micro Balance
Products
Product Category
balances (10)
Manufacturer
Weighing Capacity (g)
Package Size
Filter Products
Showing 1-10 of 10
Compare
Product Number
Description
Pricing
OH30219001
Ohaus
®
Explorer
®
Semi-Micro Balance,
Model EX225D/AD, weighing capacity 120-220 g, resolution: 0.01_mg/0.1 mg, AC/DC input 230 V, universal plug set
Expand
View Pricing
OH30139518
Ohaus
®
Explorer
®
Semi-Micro Balance,
Model EX225DM/AD, weighing capacity 120-220 g, resolution: 0.01_mg/0.1 mg, AC/DC input 230 V, universal plug set
Expand
View Pricing
OH30218999
Ohaus
®
Explorer
®
Semi-Micro Balance,
Model EX125, weighing capacity 120 g, resolution: 0.01 mg, AC/DC input 230 V, universal plug set
Expand
View Pricing
OH30139519
Ohaus
®
Explorer
®
Semi-Micro Balance,
Model EX225M/AD, weighing capacity 220 g, resolution: 0.01 mg, AC/DC input 230 V, universal plug set
Expand
View Pricing
OH30219002
Ohaus
®
Explorer
®
Semi-Micro Balance,
Model EX225/AD, weighing capacity 220 g, resolution: 0.01 mg, AC/DC input 230 V, universal plug set
Expand
View Pricing
OH30139516
Ohaus
®
Explorer
®
Semi-Micro Balance,
Model EX125M, weighing capacity 120 g, resolution: 0.01 mg, AC/DC input 230 V, universal plug set
Expand
View Pricing
OH30218998
Ohaus
®
Explorer
®
Semi-Micro Balance,
Model EX125D, weighing capacity 52_g/120 g, resolution: 0.01_mg/0.1 mg, AC/DC input 230 V, universal plug set
Expand
View Pricing
OH30139517
Ohaus
®
Explorer
®
Semi-Micro Balance,
Model EX225DM, weighing capacity 120-220 g, resolution: 0.01_mg/0.1 mg, AC/DC input 230 V, universal plug set
Expand
View Pricing
OH30219000
Ohaus
®
Explorer
®
Semi-Micro Balance,
Model EX225D, weighing capacity 120-220 g, resolution: 0.01_mg/0.1 mg, AC/DC input 230 V, universal plug set
Expand
View Pricing
OH30139515
Ohaus
®
Explorer
®
Semi-Micro Balance,
Model EX125DM, weighing capacity 52_g/120 g, resolution: 0.01_mg/0.1 mg, AC/DC input 230 V, universal plug set
Expand
View Pricing