Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Ohaus
®
Explorer
®
semi-micro analytical balance
Products
Product Category
balances (10)
Manufacturer
Diameter (mm)
Weighing Capacity (g)
Filter Products
Showing 1-10 of 10
Compare
Product Number
Description
Pricing
Z743938
Ohaus
®
Explorer
®
semi-micro analytical balance,
Model EX225M/AD, weighing capacity 220 g, Precision 0.01 mg, AC/DC input 230 V (Euro Plug and UK Plug)
Expand
View Pricing
Z743935
Ohaus
®
Explorer
®
semi-micro analytical balance,
Model EX225/AD, weighing capacity 220 g, Precision 0.01 mg, AC/DC input 230 V (Euro Plug and UK Plug)
Expand
View Pricing
Z743931
Ohaus
®
Explorer
®
semi-micro analytical balance,
Model EX225D/AD, weighing capacity 120 or 220 g, Precision 0.01 0.1 mg, AC/DC input 230 V (Euro Plug and UK Plug)
Expand
View Pricing
Z743928
Ohaus
®
Explorer
®
semi-micro analytical balance,
Model EX125, weighing capacity 120 g, Precision 0.01 mg, AC/DC input 230 V (Euro Plug and UK Plug)
Expand
View Pricing
Z743926
Ohaus
®
Explorer
®
semi-micro analytical balance,
Model EX125D, weighing capacity 51 or 120 g, Precision 0.01 0.1 mg, AC/DC input 230 V (Euro Plug and UK Plug)
Expand
View Pricing
Z743939
Ohaus
®
Explorer
®
semi-micro analytical balance,
Model EX225DM, weighing capacity 120 or 220 g, Precision 0.01 0.1 mg, AC/DC input 230 V (Euro Plug and UK Plug)
Expand
View Pricing
Z743930
Ohaus
®
Explorer
®
semi-micro analytical balance,
Model EX225D, weighing capacity 120 or 220 g, Precision 0.01 0.1 mg, AC/DC input 230 V (Euro Plug and UK Plug)
Expand
View Pricing
Z743936
Ohaus
®
Explorer
®
semi-micro analytical balance,
Model EX125M, weighing capacity 120 g, Precision 0.01 mg, AC/DC input 230 V (Euro Plug and UK Plug)
Expand
View Pricing
Z743937
Ohaus
®
Explorer
®
semi-micro analytical balance,
Model EX125DM, weighing capacity 52 or 120 g, Precision 0.01 0.1 mg, AC/DC input 230 V (Euro Plug and UK Plug)
Expand
View Pricing
Z743940
Ohaus
®
Explorer
®
semi-micro analytical balance,
Model EX225DM/AD, weighing capacity 120 g, accuracy: 220 g, Precision 0.01 0.1 mg, AC/DC input 230 V (Euro Plug and UK Plug)
Expand
View Pricing