
Ofloxacin impurity E

Synonym(s):
9-Fluoro-2,3-dihydro-3-methyl-7-oxo-10-(1-piperazinyl)-7H-pyrido[1,2,3-de]-1,4-benzoxazine-6-carboxylic acid
Empirical Formula (Hill Notation):
C17H18FN3O4
CAS Number:
Molecular Weight:
347.34
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing