Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Nunc
®
MicroWell
™
96 well polypropylene plates
Products
Product Category
plates (4)
Brand
Sigma-Aldrich (2)
Manufacturer
Binding Type
Length (mm)
Wells
Color
Plate Material
Package Size
Special Grade
Natural (4)
Sterility
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
P6866
Nunc
®
MicroWell
™
96 well polypropylene plates,
96 well plate, polypropylene, natural, round bottom, 500uL/well, non sterile, 120/cs
Expand
View Pricing
P6991
Nunc
®
MicroWell
™
96 well polypropylene plates,
96 well plate, polypropylene, natural, round bottom, 500uL/well, sterile, 120/cs
Expand
View Pricing
P7241
Nunc
®
MicroWell
™
96 well polypropylene plates,
96 well plate, polypropylene, natural, V-bottom, 500uL/well, sterile, 120/cs
Expand
View Pricing
P7116
Nunc
®
MicroWell
™
96 well polypropylene plates,
96 well plate, polypropylene, natural, V-bottom, 500uL/well, non sterile, 120/cs
Expand
View Pricing