Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Nunc
®
Lab-Tek
®
II chambered coverglass
Products
Manufacturer
Length (mm)
Surface Area cm²
Working Volume (ml)
Color
Package Size
Sterility
Filter Products
Showing 1-6 of 6
Compare
Product Number
Description
Pricing
Z734756
Nunc
®
Lab-Tek
®
II chambered coverglass,
4 wells, polystyrene chambers, 1.5 borosilicate coverglass, 1.7 cm
2
/well, 96/cs
Expand
View Pricing
Z734535
Nunc
®
Lab-Tek
®
II chambered coverglass,
1 well, polystyrene chamber, 1.5 borosilicate coverglass, 8.6 cm
2
/well, 96/cs
Expand
View Pricing
TMO155411
Nunc
®
Lab-Tek
®
II chambered coverglass,
borosilicate glass, L × W 57 mm × 25 mm, chamber size 8, sterile, case of 96 ea
Expand
View Pricing
TMO155361
Nunc
®
Lab-Tek
®
II chambered coverglass,
borosilicate glass, L × W 1 mm × 25 mm, chamber size 1, sterile, case of 96 ea
Expand
View Pricing
TMO155380
Nunc
®
Lab-Tek
®
II chambered coverglass,
borosilicate glass, L × W 57 mm × 25 mm, chamber size 2, sterile, case of 96 ea
Expand
View Pricing
TMO155383
Nunc
®
Lab-Tek
®
II chambered coverglass,
borosilicate glass, L × W 57 mm × 25 mm, chamber size 4, sterile, case of 96 ea
Expand
View Pricing