Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
NovaSeptum
®
SURe Accurate Volume Syringe Sampling Unit
Products
Product Category
syringes (4)
Brand
Millipore (4)
Product Line
Volume (ml)
Quality Segment
Application
Sterility
Sterilization Method
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
5444
NovaSeptum
®
SURe Accurate Volume Syringe Sampling Unit,
Manifold (5-valve) with Inlet C-Flex
®
Tubing, sterile; β-irradiated, volume 5 mL (20 mL)
Expand
View Pricing
AC464
NovaSeptum
®
SURe Accurate Volume Syringe Sampling Unit,
Manifold (5-valve) with AseptiQuik
®
S connector, sterile; γ-irradiated, volume 5 mL (20 mL)
Expand
View Pricing
5443
NovaSeptum
®
SURe Accurate Volume Syringe Sampling Unit,
Manifold (3-valve) with Inlet C-Flex
®
Tubing, sterile; γ-irradiated, volume 5 mL & 20 mL
Expand
View Pricing
AC463
NovaSeptum
®
SURe Accurate Volume Syringe Sampling Unit,
Manifold (3-valve) with AseptiQuik
®
S connector, sterile; γ-irridiated, volume 5 mL & 20 mL
Expand
View Pricing