Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
NovaSeptum
®
Holder CCS
Products
Product Category
holders (6)
Brand
Millipore (6)
Quality Segment
Color
Sterility
Filter Products
Showing 1-6 of 6
Compare
Product Number
Description
Pricing
AT52/5-3D0
NovaSeptum
®
Holder CCS,
Expand
View Pricing
AT12/2-3D0
NovaSeptum
®
Holder CCS,
Expand
View Pricing
AT92/6-3D0
NovaSeptum
®
Holder CCS,
Expand
View Pricing
AG22/380X252-3D0
NovaSeptum
®
Holder CCS,
Expand
View Pricing
AT72/5-3D0
NovaSeptum
®
Holder CCS,
7-port TC50,5, EP+Glass blasted, NO PTFE coating
Expand
View Pricing
AG22/380X550-3D0
NovaSeptum
®
Holder CCS,
Expand
View Pricing