Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Norell
®
Sample Vault Series
™
NMR tubes
Products
Product Category
tubes (14)
Manufacturer
Diameter (mm)
Color
Frequency (MHz)
Filter Products
Showing 1-14 of 14
Compare
Product Number
Description
Pricing
Z693189
Norell
®
Sample Vault Series
™
NMR tubes,
standard wall, closed cap, frequency 700 MHz, diam. × L 3 mm × 178 mm
Expand
View Pricing
Z693146
Norell
®
Sample Vault Series
™
NMR tubes,
standard wall, closed cap, frequency 700 MHz, diam. × L 5 mm × 178 mm
Expand
View Pricing
Z693170
Norell
®
Sample Vault Series
™
NMR tubes,
standard wall, open cap, frequency 700 MHz, diam. × L 3 mm × 103.5 mm
Expand
View Pricing
Z693278
Norell
®
Sample Vault Series
™
NMR tubes,
standard wall, closed cap, frequency 950 MHz, diam. × L 3 mm × 178 mm
Expand
View Pricing
Z693138
Norell
®
Sample Vault Series
™
NMR tubes,
standard wall, open cap, frequency 700 MHz, diam. × L 5 mm × 103.5 mm
Expand
View Pricing
Z693227
Norell
®
Sample Vault Series
™
NMR tubes,
standard wall, closed cap, frequency 950 MHz, diam. × L 5 mm × 178 mm
Expand
View Pricing
Z693243
Norell
®
Sample Vault Series
™
NMR tubes,
standard wall, closed cap, frequency 950 MHz, diam. × L 3 mm × 103.5 mm
Expand
View Pricing
Z693200
Norell
®
Sample Vault Series
™
NMR tubes,
standard wall, closed cap, frequency 950 MHz, diam. × L 5 mm × 103.5 mm
Expand
View Pricing
Z693219
Norell
®
Sample Vault Series
™
NMR tubes,
standard wall, open cap, frequency 950 MHz, diam. × L 5 mm × 103.5 mm
Expand
View Pricing
Z693154
Norell
®
Sample Vault Series
™
NMR tubes,
standard wall, open cap, frequency 700 MHz, diam. × L 5 mm × 178 mm
Expand
View Pricing
Z693286
Norell
®
Sample Vault Series
™
NMR tubes,
standard wall, open cap, frequency 950 MHz, diam. × L 3 mm × 178 mm
Expand
View Pricing
Z693251
Norell
®
Sample Vault Series
™
NMR tubes,
standard wall, open cap, frequency 950 MHz, diam. × L 3 mm × 103.5 mm
Expand
View Pricing
Z693235
Norell
®
Sample Vault Series
™
NMR tubes,
standard wall, open cap, frequency 950 MHz, diam. × L 5 mm × 178 mm
Expand
View Pricing
Z693197
Norell
®
Sample Vault Series
™
NMR tubes,
standard wall, open cap, frequency 700 MHz, diam. × L 3 mm × 178 mm
Expand
View Pricing