Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
N,N,N′,N′
-Tetramethyl-
p
-phenylenediamine dihydrochloride
Synonym(s):
TMPPD, Wurster’s reagent
Linear Formula:
C
6
H
4
[N(CH
3
)
2
]
2
·2HCl
CAS Number:
637-01-4
Molecular Weight:
237.17
EC Number:
211-274-8
Products
Product Category
building blocks (1)
substrates (1)
Brand
Sigma-Aldrich (1)
Color
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Application
Markush Class
Markush Group
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
T3134
N,N,N′,N′
-Tetramethyl-
p
-phenylenediamine dihydrochloride,
≥95%, powder
Expand
View Pricing
87890
N,N,N′,N′
-Tetramethyl-
p
-phenylenediamine dihydrochloride,
≥97.0% (AT)
Expand
View Pricing