Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
N
,
N
,
N
′,
N
′-Tetramethylethylenediamine
Synonym(s):
1,2-Bis(dimethylamino)ethane, TEMED, TMEDA, N,N,N′,N′-Tetramethyl ethylenediamine
Linear Formula:
(CH
3
)
2
NCH
2
CH
2
N(CH
3
)
2
CAS Number:
110-18-9
Molecular Weight:
116.20
EC Number:
203-744-6
Products
Product Category
amines (1)
building blocks (1)
Brand
Sigma-Aldrich (1)
Boiling Point (°C)
Feature
Formula Weight
Physical Form
Product Line
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Technique
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
T9281
N
,
N
,
N
′,
N
′-Tetramethylethylenediamine,
BioReagent, suitable for electrophoresis, ≥99.0%
Expand
View Pricing
T7024
N
,
N
,
N
′,
N
′-Tetramethylethylenediamine,
BioReagent, Molecular Biology, ≥99% (GC)
Expand
View Pricing
411019
N
,
N
,
N
′,
N
′-Tetramethylethylenediamine,
≥99.5%, purified by redistillation
Expand
View Pricing
8.08742
N
,
N
,
N
′,
N
′-Tetramethylethylenediamine,
for synthesis
Expand
View Pricing
T22500
N
,
N
,
N
′,
N
′-Tetramethylethylenediamine,
ReagentPlus
®
, 99%
Expand
View Pricing