Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
NMR tube caps
Products
Product Category
caps (11)
Brand
Sigma-Aldrich (6)
Color
Manufacturer
Filter Products
Showing 1-11 of 11
Compare
Product Number
Description
Pricing
Z118079
NMR tube caps,
NMR tube size 5 mm
Expand
View Pricing
Z153281
NMR tube caps,
NMR tube size 5 mm
Expand
View Pricing
Z153273
NMR tube caps,
NMR tube size 5 mm
Expand
View Pricing
Z153303
NMR tube caps,
NMR tube size 5 mm
Expand
View Pricing
Z153311
NMR tube caps,
NMR tube size 5 mm
Expand
View Pricing
Z118087
NMR tube caps,
NMR tube size 10 mm
Expand
View Pricing
Z153338
NMR tube caps,
NMR tube size 5 mm
Expand
View Pricing
Z153265
NMR tube caps,
NMR tube size 5 mm, assorted colors
Expand
View Pricing
Z272078
NMR tube caps,
NMR tube size 3 mm
Expand
View Pricing
Z567485
NMR tube caps,
NMR tube size 4 mm, blue
Expand
View Pricing
Z567477
NMR tube caps,
NMR tube size 2 mm, red
Expand
View Pricing