Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
N
-Methyl-D-glucamine
Synonym(s):
1-Deoxy-1-(methylamino)-D-glucitol, Meglumine
Empirical Formula (Hill Notation):
C
7
H
17
NO
5
CAS Number:
6284-40-8
Molecular Weight:
195.21
EC Number:
228-506-9
Products
Product Category
building blocks (1)
carbohydrates (1)
Brand
Sigma-Aldrich (2)
Biological Source
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Quality Segment
Markush Class
Markush Group
Optical Isomer
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
M2004
N
-Methyl-D-glucamine,
99.0-100.5% (titration)
Expand
View Pricing
66930
N
-Methyl-D-glucamine,
ReagentPlus
®
, ≥99.0% (T)
Expand
View Pricing