Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Nitinol alloy, Ni55Ti45
Products
Manufacturer
Physical Form
Diameter (mm)
ID (mm)
Length (m)
Length (mm)
OD (mm)
Filter Products
Showing 1-16 of 16
Compare
Product Number
Description
Pricing
GF32642669
Nitinol alloy, Ni55Ti45,
5m wire, 0.8mm diameter, shape-memory alloy
Expand
View Pricing
GF20375300
Nitinol alloy, Ni55Ti45,
0.5m wire, 0.8mm diameter, shape-memory alloy
Expand
View Pricing
GF43925924
Nitinol alloy, Ni55Ti45,
1m wire, 0.8mm diameter, shape-memory alloy
Expand
View Pricing
GF79186348
Nitinol alloy, Ni55Ti45,
10m wire, 0.8mm diameter, shape-memory alloy
Expand
View Pricing
GF98944958
Nitinol alloy, Ni55Ti45,
2m wire, 0.8mm diameter, shape-memory alloy
Expand
View Pricing
GF83853456
Nitinol alloy, Ni55Ti45,
200mm tube, 0.8mm outside diameter, 0.1mm wall thickness, 0.6mm inside diameter, superelastic
Expand
View Pricing
GF94363982
Nitinol alloy, Ni55Ti45,
100mm tube, 1.27mm outside diameter, 0.15mm wall thickness, 0.97mm inside diameter, superelastic
Expand
View Pricing
GF55165664
Nitinol alloy, Ni55Ti45,
500mm tube, 0.8mm outside diameter, 0.1mm wall thickness, 0.6mm inside diameter, superelastic
Expand
View Pricing
GF47582017
Nitinol alloy, Ni55Ti45,
500mm tube, 1.27mm outside diameter, 0.15mm wall thickness, 0.97mm inside diameter, superelastic
Expand
View Pricing
GF30522484
Nitinol alloy, Ni55Ti45,
1000mm tube, 0.8mm outside diameter, 0.1mm wall thickness, 0.6mm inside diameter, superelastic
Expand
View Pricing
GF62371199
Nitinol alloy, Ni55Ti45,
100mm tube, 1.8mm outside diameter, 0.125mm wall thickness, 1.55mm inside diameter, superelastic
Expand
View Pricing
GF42816135
Nitinol alloy, Ni55Ti45,
500mm tube, 1.8mm outside diameter, 0.125mm wall thickness, 1.55mm inside diameter, superelastic
Expand
View Pricing
GF02714767
Nitinol alloy, Ni55Ti45,
200mm tube, 1.8mm outside diameter, 0.125mm wall thickness, 1.55mm inside diameter, superelastic
Expand
View Pricing
GF93667843
Nitinol alloy, Ni55Ti45,
200mm tube, 1.27mm outside diameter, 0.15mm wall thickness, 0.97mm inside diameter, superelastic
Expand
View Pricing
GF88255659
Nitinol alloy, Ni55Ti45,
1000mm tube, 1.8mm outside diameter, 0.125mm wall thickness, 1.55mm inside diameter, superelastic
Expand
View Pricing
GF97831996
Nitinol alloy, Ni55Ti45,
1000mm tube, 1.27mm outside diameter, 0.15mm wall thickness, 0.97mm inside diameter, superelastic
Expand
View Pricing