Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Nickel oxide
Products
Color
Manufacturer
Physical Form
Particle Size (µm)
Filter Products
Showing 1-9 of 9
Compare
Product Number
Description
Pricing
GF37536396
Nickel oxide,
powder, max. particle size 45 micron, weight 100 g, green
Expand
View Pricing
GF30173420
Nickel oxide,
powder, max. particle size 500 micron, weight 25 g, min. particle size 200 micron
Expand
View Pricing
GF22948094
Nickel oxide,
powder, max. particle size 45 micron, weight 100 g, black
Expand
View Pricing
GF63811795
Nickel oxide,
powder, 45 max. part. size (micron), weight 50 g, black
Expand
View Pricing
GF54705966
Nickel oxide,
powder, max. particle size 1700 micron, weight 25 g, min. particle size 850 micron
Expand
View Pricing
GF83449064
Nickel oxide,
powder, 45 max. part. size (micron), weight 200 g, black
Expand
View Pricing
GF17641346
Nickel oxide,
powder, max. particle size 45 micron, weight 50 g, green
Expand
View Pricing
GF28835736
Nickel oxide,
powder, max. particle size 45 micron, weight 500 g, green
Expand
View Pricing
GF87977952
Nickel oxide,
powder, 45 max. part. size (micron), weight 500 g, black
Expand
View Pricing