Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Nickel/Iron/Molybdenum
Products
Manufacturer
Physical Form
Length (m)
Filter Products
Showing 1-6 of 6
Compare
Product Number
Description
Pricing
GF66534958
Nickel/Iron/Molybdenum,
wire, Ni80%/Fe15.5%/Mo 4.5%, length 100 m, 0.5 mm diameter, temper as drawn
Expand
View Pricing
GF14168033
Nickel/Iron/Molybdenum,
wire, Ni 80%/Fe15.5%/Mo 4.5, 0.5 mm diameter, length 50 m, temper as drawn
Expand
View Pricing
GF06772057
Nickel/Iron/Molybdenum,
wire, Ni 80%/Fe15.5%/Mo 4.5, 1.0 mm diameter, length 20 m, temper as drawn
Expand
View Pricing
GF02413011
Nickel/Iron/Molybdenum,
wire, Ni 80%/Fe15.5%/Mo 4.5, 1.0 mm diameter, length 50 m, temper as drawn
Expand
View Pricing
GF14561478
Nickel/Iron/Molybdenum,
wire, Ni 80%/Fe15.5%/Mo 4.5, 1.0 mm diameter, length 10 m, temper as drawn
Expand
View Pricing
GF39859048
Nickel/Iron/Molybdenum,
wire, Ni 80%/Fe15.5%/Mo 4.5, 1.0 mm diameter, length 1 m, temper as drawn
Expand
View Pricing