Nickel(II) phthalocyanine-tetrasulfonic acid tetrasodium salt
Synonym(s):
Phthalocyanine-tetrasulfonic acid Nickel(II) complex tetrasodium salt
Empirical Formula (Hill Notation):
C32H12N8Na4NiO12S4
CAS Number:
Molecular Weight:
979.40
Products
Brand
Formula Weight
Quality Segment
Reagent Type
Core
Showing 1-1 of 1
Compare
Product Number
Description
Pricing