Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Nickel-chromium alloy, Ni80Cr20
Products
Manufacturer
Physical Form
Diameter (mm)
Length (m)
Filter Products
Showing 1-20 of 201
Page 1 of 11
Page 1 of 11
Compare
Product Number
Description
Pricing
GF70959796
Nickel-chromium alloy, Ni80Cr20,
100m wire, 0.125mm diameter, hard
Expand
View Pricing
GF10092990
Nickel-chromium alloy, Ni80Cr20,
100m wire, 0.5mm diameter, annealed
Expand
View Pricing
GF28737322
Nickel-chromium alloy, Ni80Cr20,
5m wire, 0.01mm diameter, as drawn
Expand
View Pricing
GF91593478
Nickel-chromium alloy, Ni80Cr20,
foil, 1m coil, 2.5mm coil width, 0.50mm thickness, annealed
Expand
View Pricing
GF34941438
Nickel-chromium alloy, Ni80Cr20,
200mm rod, 3.0mm diameter
Expand
View Pricing
GF15510563
Nickel-chromium alloy, Ni80Cr20,
foil, 100 x 100mm, 0.025mm thickness, as rolled, light tight (lt)
Expand
View Pricing
GF92803653
Nickel-chromium alloy, Ni80Cr20,
powder, 75micron max. particle size, 100g
Expand
View Pricing
GF91818265
Nickel-chromium alloy, Ni80Cr20,
1m wire, 0.020mm diameter, as drawn
Expand
View Pricing
GF74075496
Nickel-chromium alloy, Ni80Cr20,
1000mm rod, 3.0mm diameter
Expand
View Pricing
GF47331783
Nickel-chromium alloy, Ni80Cr20,
200mm tube, 1.3mm outside diameter, 0.25mm wall thickness, 0.8mm inside diameter
Expand
View Pricing
GF22979526
Nickel-chromium alloy, Ni80Cr20,
foil, 1m coil, 1mm coil width, 0.20mm thickness, annealed
Expand
View Pricing
GF94253205
Nickel-chromium alloy, Ni80Cr20,
250m wire, 0.75mm diameter, annealed
Expand
View Pricing
GF95046240
Nickel-chromium alloy, Ni80Cr20,
50m wire, 0.25mm diameter, hard
Expand
View Pricing
GF54020540
Nickel-chromium alloy, Ni80Cr20,
25m wire, 0.125mm diameter, hard
Expand
View Pricing
GF89552201
Nickel-chromium alloy, Ni80Cr20,
foil, 100 x 100mm, 0.25mm thickness, as rolled
Expand
View Pricing
GF30537121
Nickel-chromium alloy, Ni80Cr20,
100mm rod, 3.0mm diameter
Expand
View Pricing
GF01378290
Nickel-chromium alloy, Ni80Cr20,
200mm rod, 6.3mm diameter
Expand
View Pricing
GF98184606
Nickel-chromium alloy, Ni80Cr20,
5m wire, 0.015mm diameter, as drawn
Expand
View Pricing
GF01651115
Nickel-chromium alloy, Ni80Cr20,
foil, 100 x 100mm, 0.18mm thickness, as rolled
Expand
View Pricing
GF60682518
Nickel-chromium alloy, Ni80Cr20,
foil, 200 x 200mm, 0.10mm thickness, as rolled
Expand
View Pricing
Page 1 of 11
Page 1 of 11