Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
n-Heptane
Synonym(s):
Heptane
Linear Formula:
CH
3
(CH
2
)
5
CH
3
CAS Number:
142-82-5
Molecular Weight:
100.20
Products
Product Category
solvents (7)
Brand
Supelco (7)
Sustainability
Boiling Point (°C)
Feature
Formula Weight
Physical Form
Product Line
Quality Segment
Technique
Filter Products
Showing 1-7 of 7
Compare
Product Number
Description
Pricing
1.04379
n-Heptane,
for analysis EMSURE
®
Reag. Ph Eur
Expand
View Pricing
1.04390
n-Heptane,
for liquid chromatography LiChrosolv
®
Expand
View Pricing
1.04365
n-Heptane,
EMPLURA
®
Expand
View Pricing
1.04360
n-Heptane,
for gas chromatography ECD and FID SupraSolv
®
Expand
View Pricing
1.03654
n-Heptane,
hypergrade for LC-MS LiChrosolv
®
Expand
View Pricing
1.04366
n-Heptane,
for spectroscopy Uvasol
®
Expand
View Pricing
1.04307
n-Heptane,
about 85% EMPLURA
®
Expand
View Pricing