
N-Boc-trans-4-hydroxy-L-proline methyl ester

Empirical Formula (Hill Notation):
C11H19NO5
CAS Number:
Molecular Weight:
245.27
Products
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Application
Markush Class
Markush Group
Optical Isomer
Amino Acid
Amino Acid Type
Reaction Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing