
Naringenin

Synonym(s):
(±)-Naringenin, (±)-2,3-Dihydro-5,7-dihydroxy-2-(4-hydroxyphenyl)-4H-1-benzopyran-4-one, 4′,5,7-Trihydroxyflavanone
Empirical Formula (Hill Notation):
C15H12O5
CAS Number:
Molecular Weight:
272.25
Products
Product Category
Brand
Biological Source
Formula Weight
Manufacturer
Melting Point (°C)
Product Line
Purity
Format
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing