Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Nalgene
®
white caps
Products
Product Category
caps (5)
Manufacturer
Diameter (mm)
Color
Package Size
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
TMO362150-1384
Nalgene
®
white caps,
polypropylene, diam. × thread 41 mm × 38-430 mm, case of 250 ea
Expand
View Pricing
TMO362150-1280
Nalgene
®
white caps,
polypropylene, diam. × thread 30 mm × 28-415 mm, case of 500 ea
Expand
View Pricing
TMO362150-1200
Nalgene
®
white caps,
polypropylene, diam. × thread 22 mm × 20-415 mm, case of 2000 ea
Expand
View Pricing
TMO362150-1130
Nalgene
®
white caps,
polypropylene, diam. × thread 16 mm × 13-415 mm, case of 2000 ea
Expand
View Pricing
TMO362150-1240
Nalgene
®
white caps,
polypropylene, diam. × thread 27 mm × 24-415 mm, case of 1000 ea
Expand
View Pricing