Skip to Content
Products
Cart
73
AD
EN
Products
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Login
Order Lookup
Quick Order
Cart
73
Back
Nalgene
®
narrow-mouth bottle without cap
Products
Product Category
bottles (9)
Manufacturer
Capacity (mL)
Volume (ml)
Color
Package Size
Special Grade
Natural (6)
Filter Products
Showing 1-9 of 9
Compare
Product Number
Description
Pricing
TMO362089-0032
Nalgene
®
narrow-mouth bottle without cap,
natural high-density polyethylene, volume 1000 mL, case of 50 ea
Expand
View Pricing
TMO362006-9050
Nalgene
®
narrow-mouth bottle without cap,
amber polypropylene copolymer, volume 15 mL, case of 2000 ea
Expand
View Pricing
TMO362089-0001
Nalgene
®
narrow-mouth bottle without cap,
natural high-density polyethylene, volume 30 mL, case of 1000 ea
Expand
View Pricing
TMO362089-0016
Nalgene
®
narrow-mouth bottle without cap,
natural high-density polyethylene, volume 500 mL, case of 125 ea
Expand
View Pricing
TMO362006-9125
Nalgene
®
narrow-mouth bottle without cap,
amber polypropylene copolymer, volume 4 mL, case of 2000 ea
Expand
View Pricing
TMO362006-9025
Nalgene
®
narrow-mouth bottle without cap,
amber polypropylene copolymer, volume 8 mL, case of 2000 ea
Expand
View Pricing
TMO362089-0008
Nalgene
®
narrow-mouth bottle without cap,
natural high-density polyethylene, volume 250 mL, case of 250 ea
Expand
View Pricing
TMO362089-0002
Nalgene
®
narrow-mouth bottle without cap,
natural high-density polyethylene, volume 60 mL, case of 1000 ea
Expand
View Pricing
TMO362089-0004
Nalgene
®
narrow-mouth bottle without cap,
natural high-density polyethylene, volume 125 mL, case of 500 ea
Expand
View Pricing
Choose up to 4 products to compare
Compare
Clear All
English - EN
Español - ES
Français - FR
Learn More