Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Nalgene
®
Griffin beakers
Products
Product Category
beakers (13)
Brand
Sigma-Aldrich (1)
Manufacturer
Capacity (mL)
Volume (ml)
Color
Package Size
Filter Products
Showing 1-13 of 13
Compare
Product Number
Description
Pricing
Z130982
Nalgene
®
Griffin beakers,
capacity 1,000 mL, translucent polypropylene
Expand
View Pricing
Z380377
Nalgene
®
Griffin beakers,
capacity 2,000 mL, translucent polypropylene
Expand
View Pricing
Z130923
Nalgene
®
Griffin beakers,
capacity 1,000 mL, transparent polymethylpentene
Expand
View Pricing
TMO1201-4000
Nalgene
®
Griffin beakers,
polypropylene, volume 4000 mL, case of 4 ea
Expand
View Pricing
Z130915
Nalgene
®
Griffin beakers,
capacity 250 mL, transparent polymethylpentene
Expand
View Pricing
Z130958
Nalgene
®
Griffin beakers,
capacity 100 mL, translucent polypropylene
Expand
View Pricing
Z130974
Nalgene
®
Griffin beakers,
capacity 250 mL, translucent polypropylene
Expand
View Pricing
Z130966
Nalgene
®
Griffin beakers,
capacity 150 mL, translucent polypropylene
Expand
View Pricing
Z130931
Nalgene
®
Griffin beakers,
capacity 50 mL, translucent polypropylene
Expand
View Pricing
Z130893
Nalgene
®
Griffin beakers,
capacity 100 mL, transparent polymethylpentene
Expand
View Pricing
Z130907
Nalgene
®
Griffin beakers,
capacity 150 mL, transparent polymethylpentene
Expand
View Pricing
Z130885
Nalgene
®
Griffin beakers,
capacity 50 mL, transparent polymethylpentene
Expand
View Pricing
TMO1201-0030
Nalgene
®
Griffin beakers,
polypropylene, volume 30 mL, case of 18 ea
Expand
View Pricing