Skip to Content
Merck
Nalgene® Cryoware marker set assorted colors, one each red, green, blue, and black

Nalgene® Cryoware marker set

Nalgene® Cryoware marker set assorted colors, one each red, green, blue, and black

Products

Color
Manufacturer
Package Size
Technique
Showing 1-2 of 2