Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Nalgene
®
Certified Clean bottles
Products
Product Category
bottles (7)
Manufacturer
Capacity (mL)
Volume (ml)
Color
Package Size
Filter Products
Showing 1-7 of 7
Compare
Product Number
Description
Pricing
TMO382019-0030
Nalgene
®
Certified Clean bottles,
clear PETG, volume 30 mL, case of 72 ea
Expand
View Pricing
TMO382019-0500
Nalgene
®
Certified Clean bottles,
clear PETG, volume 500 mL, case of 24 ea
Expand
View Pricing
TMO382019-0125
Nalgene
®
Certified Clean bottles,
clear PETG, volume 125 mL, case of 48 ea
Expand
View Pricing
TMO382019-1000
Nalgene
®
Certified Clean bottles,
clear PETG, volume 1000 mL, case of 24 ea
Expand
View Pricing
TMO382019-0250
Nalgene
®
Certified Clean bottles,
clear PETG, volume (250 mL), case of 48 ea
Expand
View Pricing
TMO382019-2000
Nalgene
®
Certified Clean bottles,
clear PETG, volume 2000 mL, case of 12 ea
Expand
View Pricing
TMO382019-0060
Nalgene
®
Certified Clean bottles,
clear PETG, volume 60 mL, case of 72 ea
Expand
View Pricing