![NAD Grade II, free acid](/deepweb/assets/sigmaaldrich/product/structures/346/044/40a29c82-f86a-4973-8389-14b5aed87785/640/40a29c82-f86a-4973-8389-14b5aed87785.png)
NAD
![NAD Grade II, free acid](/deepweb/assets/sigmaaldrich/product/structures/346/044/40a29c82-f86a-4973-8389-14b5aed87785/640/40a29c82-f86a-4973-8389-14b5aed87785.png)
Synonym(s):
β-Nicotinamide adenine dinucleotide hydrate, β-DPN, β-NAD, Coenzyme 1, Cozymase, DPN, Diphosphopyridine nucleotide, NAD, Nadide
Empirical Formula (Hill Notation):
C21H27N7O14P2 · xH2O
CAS Number:
Molecular Weight:
663.43 (anhydrous basis)
Products
Color
Formula Weight
Manufacturer
pH Value
Physical Form
Showing 1-2 of 2
Compare
Product Number
Description
Pricing