
N-Acetyl-L-tryptophan

Empirical Formula (Hill Notation):
C13H14N2O3
CAS Number:
Molecular Weight:
246.26
Products
Product Category
Brand
Color
Formula Weight
Physical Form
Agency
Format
Quality Segment
Application
Optical Isomer
Amino Acid
API Family
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2