Skip to Content
Merck
N-Acetyl-D-lactosamine ≥98% (TLC)

N-Acetyl-D-lactosamine

N-Acetyl-D-lactosamine ≥98% (TLC)
Synonym(s):
β-D-Gal-(1→4)-D-GlcNAc, 2-Acetamido-2-deoxy-4-O-β-D-galactopyranosyl-D-glucopyranose, LN, N-Acetyl-4-O-(β-D-galactopyranosyl)-D-glucosamine
Empirical Formula (Hill Notation):
C14H25NO11
CAS Number:
Molecular Weight:
383.35

Products

Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Quality Segment
Optical Isomer
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing