Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
N
-(2-Hydroxyethyl)ethylenediamine-
N
,
N
′,
N
′-triacetic acid
Synonym(s):
N
-Carboxymethyl-
N′
-(2-hydroxyethyl)-
N,N′
-ethylenediglycine, HEDTA, HEEDTA
Linear Formula:
HOCH
2
CH
2
N(CH
2
CO
2
H)CH
2
CH
2
N(CH
2
CO
2
H)
2
CAS Number:
150-39-0
Molecular Weight:
278.26
EC Number:
205-759-3
Products
Brand
Sigma-Aldrich (2)
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Quality Segment
Markush Class
Markush Group
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
H8126
N
-(2-Hydroxyethyl)ethylenediamine-
N
,
N
′,
N
′-triacetic acid,
~98%
Expand
View Pricing
H7154
N
-(2-Hydroxyethyl)ethylenediamine-
N
,
N
′,
N
′-triacetic acid,
BioXtra, ≥98%
Expand
View Pricing