Skip to Content
Merck
N-(2-Aminoethyl)maleimide trifluoroacetate salt ≥95% (HPLC), ≥98% (T)

N-(2-Aminoethyl)maleimide trifluoroacetate salt

N-(2-Aminoethyl)maleimide trifluoroacetate salt ≥95% (HPLC), ≥98% (T)
Synonym(s):
2-Maleimidoethylamine trifluoroacetate salt
Empirical Formula (Hill Notation):
C6H8N2O2 · C2HF3O2
CAS Number:
Molecular Weight:
254.16

Products

Formula Weight
Physical Form
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Functionality
Reagent Type
Special Grade
Showing 1-1 of 1