Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Myoglobin from equine heart
Synonym(s):
Myoglobin from horse heart
CAS Number:
100684-32-0
EC Number:
309-705-0
Products
Product Category
proteins (2)
Brand
Sigma-Aldrich (1)
Supelco (1)
Biological Source
Physical Form
pK Value
Purity
Tissue
Quality Segment
Technique
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
M1882
Myoglobin from equine heart,
≥90% (SDS-PAGE), essentially salt-free, lyophilized powder
Expand
View Pricing
M9267
Myoglobin from equine heart,
Isoelectric focusing marker, pI (1) 6.8, (2) 7.2
Expand
View Pricing