Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Multi-Parameter Refractive Index & Density Certified Reference Material at 15 °C, 20 °C and 25 °C, Single 30 mL
EC Number:
201-965-2
Products
Manufacturer
Application
Reference Material Type
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
PSRVD01
Multi-Parameter Refractive Index & Density Certified Reference Material at 15 °C, 20 °C and 25 °C, Single 30 mL,
Nominal RI Value 1.3330 at 20 °C
Expand
View Pricing
PSRVD03
Multi-Parameter Refractive Index & Density Certified Reference Material at 15 °C, 20 °C and 25 °C, Single 30 mL,
Nominal RI Value 1.5463 at 20 °C
Expand
View Pricing
PSRVD02
Multi-Parameter Refractive Index & Density Certified Reference Material at 15 °C, 20 °C and 25 °C, Single 30 mL,
Nominal RI Value 1.4217 at 20 °C
Expand
View Pricing
PSRVD04
Multi-Parameter Refractive Index & Density Certified Reference Material at 15 °C, 20 °C and 25 °C, Single 30 mL,
Nominal RI Value 1.6579 at 20 °C
Expand
View Pricing