Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
MTC
™
Bio Western Blot Box
Products
Manufacturer
Color
Package Size
Filter Products
Showing 1-12 of 12
Compare
Product Number
Description
Pricing
MTCB12007BK
MTC
™
Bio Western Blot Box,
L × W × D 3 1/2 in. (8.9 cm) × 2 9/16 in. (6.5 cm) × 1 in. (2.5 cm), black
Expand
View Pricing
MTCB120013BK
MTC
™
Bio Western Blot Box,
L × W × D 4 5/8 in. (11.7 cm) × 3 1/2 in. (8.9 cm) × 1 1/8 in. (2.8 cm), black
Expand
View Pricing
MTCB120019
MTC
™
Bio Western Blot Box,
L × W × D 6 in. (15.2 cm) × 4 in. (10.2 cm) × 2 in. (5.1 cm), clear
Expand
View Pricing
MTCB12003
MTC
™
Bio Western Blot Box,
L × W × D 2 7/8 in. (7.3 cm) × 1 3/16 in. (3 cm) × 3/4 in. (1.9 cm), clear
Expand
View Pricing
MTCB120017
MTC
™
Bio Western Blot Box,
L × W × D 6 in. (15.2 cm) × 4 in. (10.2 cm) × 1 1/4 in. (3.2 cm), clear
Expand
View Pricing
MTCB120011
MTC
™
Bio Western Blot Box,
L × W × D 4 3/8 in. (11.1 cm) × 2 1/16 in. (5.2 cm) × 1 7/8 in. (4.7 cm), clear
Expand
View Pricing
MTCB120015BK
MTC
™
Bio Western Blot Box,
L × W × D 4 9/16 in. (11.6 cm) × 4 9/16 in. (11.6 cm) × 1 1/4 in. (3.2 cm), black
Expand
View Pricing
MTCB12005
MTC
™
Bio Western Blot Box,
L × W × D 2 7/8 in. (7.3 cm) × 2 in. (5.1 cm) × 1 1/4 in. (3.2 cm), clear
Expand
View Pricing
MTCB120013
MTC
™
Bio Western Blot Box,
L × W × D 4 5/8 in. (11.7 cm) × 3 1/2 in. (8.9 cm) × 1 1/8 in. (2.8 cm), clear
Expand
View Pricing
MTCB12009
MTC
™
Bio Western Blot Box,
L × W × D 3 3/4 in. (9.5 cm) × 1 3/16 in. (3 cm) × 5/8 in. (1.6 cm), clear
Expand
View Pricing
MTCB12007
MTC
™
Bio Western Blot Box,
L × W × D 3 1/2 in. (8.9 cm) × 2 9/16 in. (6.5 cm) × 1 in. (2.5 cm), clear
Expand
View Pricing
MTCB120015
MTC
™
Bio Western Blot Box,
L × W × D 4 9/16 in. (11.6 cm) × 4 9/16 in. (11.6 cm) × 1 1/4 in. (3.2 cm), clear
Expand
View Pricing