Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
MTC
™
Bio Screw Cap Centrifuge Tube
Products
Product Category
tubes (12)
Manufacturer
Capacity (mL)
Diameter (mm)
Color
Package Size
Sterility
Filter Products
Showing 1-12 of 12
Compare
Product Number
Description
Pricing
MTCC2410
MTC
™
Bio Screw Cap Centrifuge Tube,
capacity 10 mL, sterile
Expand
View Pricing
MTCC2625R
MTC
™
Bio Screw Cap Centrifuge Tube,
capacity 25 mL, conical bottom, sterile, pack of 200 ea (8 x foam racks 25 ea)
Expand
View Pricing
MTCC2625B
MTC
™
Bio Screw Cap Centrifuge Tube,
capacity 25 mL, conical bottom, sterile, pack of 200 ea (8 x bags 25ea)
Expand
View Pricing
MTCC2600
MTC
™
Bio Screw Cap Centrifuge Tube,
capacity 15 mL, conical bottom, non-sterile, pack of 500 ea (10 x foam racks 50 ea)
Expand
View Pricing
MTCC2605
MTC
™
Bio Screw Cap Centrifuge Tube,
capacity 50 mL, sterile, pack of 500 ea (20 x bags 25 ea)
Expand
View Pricing
MTCC2603B
MTC
™
Bio Screw Cap Centrifuge Tube,
capacity 50 mL, conical bottom, non-sterile, pack of 500 ea (20 x bags 25 ea)
Expand
View Pricing
MTCC2615W
MTC
™
Bio Screw Cap Centrifuge Tube,
capacity 15 mL, conical bottom, sterile, pkg of 300 ea (individually wrapped)
Expand
View Pricing
MTCC2601
MTC
™
Bio Screw Cap Centrifuge Tube,
capacity 15 mL, conical bottom, sterile, pack of 500 ea (10 x bags 50 ea)
Expand
View Pricing
MTCC2650W
MTC
™
Bio Screw Cap Centrifuge Tube,
capacity 50 mL, conical bottom, sterile, pkg of 300 ea (individually wrapped)
Expand
View Pricing
MTCC2602
MTC
™
Bio Screw Cap Centrifuge Tube,
capacity 50 mL, conical bottom, non-sterile, pack of 500 ea (20 x foam racks 25 ea)
Expand
View Pricing
MTCC2630
MTC
™
Bio Screw Cap Centrifuge Tube,
capacity 50 mL, sterile, pack of 500 ea (20 x bags 25 ea)
Expand
View Pricing
MTCC2603
MTC
™
Bio Screw Cap Centrifuge Tube,
capacity 50 mL, conical bottom, non-sterile, pkg of 500 ea (bulk bag)
Expand
View Pricing