Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
MTC
™
Bio ClearSeal
™
Graduated Screw Cap Microtubes
Products
Product Category
tubes (4)
Manufacturer
Capacity (mL)
Diameter (mm)
Cap
Color
Package Size
Sterility
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
MTCC3220SG
MTC
™
Bio ClearSeal
™
Graduated Screw Cap Microtubes,
capacity 2.0 mL, sterile, cap, natural, self standing, pack of 1000 ea (20 x bags 50 ea)
Expand
View Pricing
MTCC3220CG
MTC
™
Bio ClearSeal
™
Graduated Screw Cap Microtubes,
capacity 2.0 mL, sterile, cap, natural, conical bottom, pack of 1000 ea (20 x bags 50 ea)
Expand
View Pricing
MTCC3215SG
MTC
™
Bio ClearSeal
™
Graduated Screw Cap Microtubes,
capacity 1.5 mL, sterile, cap, natural, self standing, pack of 1000 ea (20 x bags 50 ea)
Expand
View Pricing
MTCC3220SGL
MTC
™
Bio ClearSeal
™
Graduated Screw Cap Microtubes,
capacity 2.0 mL, sterile, cap, natural(looped), self standing, pack of 1000 ea (20 x bags 50 ea)
Expand
View Pricing