Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
mPAGE
®
Trial Kit
Products
Product Category
kits (6)
Brand
Millipore (6)
Filter Products
Showing 1-6 of 6
Compare
Product Number
Description
Pricing
MP41G12TR1
mPAGE
®
Trial Kit,
includes 2 4-12% Bis-Tris precast gels, 10x8 cm, 12-well, MOPS SDS powder, makes 1L of running buffer, 2 tank adapters
Expand
View Pricing
MP42G12TR1
mPAGE
®
Trial Kit,
includes two 4-20% Bis-Tris precast gels, 10x8 cm, 12-well, MOPS SDS powder, makes 1L of running buffer, 2 tank adapters
Expand
View Pricing
MP41G12TR2
mPAGE
®
Trial Kit,
includes two 4-12% Bis-Tris precast gels, 10x8 cm, 12-well, MES SDS powder, makes 1L of running buffer, 2 tank adapters
Expand
View Pricing
MP10W12TR1
mPAGE
®
Trial Kit,
includes two 10% Bis-Tris precast gels, 10x8 cm, 12-well, MOPS SDS powder, makes 1L of running buffer, 2 tank adapters
Expand
View Pricing
MP12W12TR2
mPAGE
®
Trial Kit,
includes two 12% Bis-Tris precast gels, 10x8 cm, 12-well, MES SDS powder, makes 1L of running buffer, 2 tank adapters
Expand
View Pricing
MP10W12TR2
mPAGE
®
Trial Kit,
includes two 10% Bis-Tris precast gels, 10x8 cm, 12-well, MES SDS powder, makes 1L of running buffer, 2 tank adapters
Expand
View Pricing