
Monoclonal Anti-Human IgG (Fc specific) antibody produced in mouse

Synonym(s):
Monoclonal Anti-Human IgG (Fc specific)
Products
Product Category
Brand
Biological Source
Clonality
Isotype
Antibody Form
Research Area
Conjugate
Quality Segment
Post Translational Modification
Species Reactivity
Technique
Showing 1-2 of 2