
Midodrine hydrochloride

Synonym(s):
(±)-1-(2,5-Dimethoxyphenyl)-2-glycinamidoethanol, Midodrine hydrochloride
Empirical Formula (Hill Notation):
C12H18N2O4 · HCl
CAS Number:
Molecular Weight:
290.74
Products
Brand
Formula Weight
Gene Alias
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Technique
Showing 1-3 of 3