Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Microtube Storage Box
Products
Product Category
boxes (7)
Manufacturer
Diameter (mm)
Wells
Color
Package Size
Special Grade
Natural (1)
Filter Products
Showing 1-7 of 7
Compare
Product Number
Description
Pricing
HS120201
Microtube Storage Box,
blue polypropylene, pk of 5, Holds 100 x 1.5-2.0 mL tubes
Expand
View Pricing
HS120044
Microtube Storage Box,
Holds 100 x 1.5-2.0 mL tubes, assorted colors polypropylene ((blue, green, purple, yellow, orange), pk of 5
Expand
View Pricing
HS120203
Microtube Storage Box,
yellow polypropylene, pk of 5, Holds 100 x 1.5-2.0 mL tubes
Expand
View Pricing
HS120042
Microtube Storage Box,
Holds 100 x 1.5-2.0 mL tubes, size 100 wells, natural polypropylene, pk of 5
Expand
View Pricing
HS120043
Microtube Storage Box,
Holds 100 x 1.5-2.0 mL tubes, black polypropylene, pk of 5
Expand
View Pricing
HS120202
Microtube Storage Box,
green polypropylene, pk of 5, Holds 100 x 1.5-2.0 mL tubes
Expand
View Pricing
HS120204
Microtube Storage Box,
orange polypropylene, pk of 5, Holds 100 x 1.5-2.0 mL tubes
Expand
View Pricing