Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Microsomes from Liver, Pooled
Products
Product Category
enzymes (3)
peptides (2)
Brand
Sigma-Aldrich (6)
Biological Source
Gene Alias
Physical Form
Quality Segment
Package Size
Filter Products
Showing 1-6 of 6
Compare
Product Number
Description
Pricing
M0317
Microsomes from Liver, Pooled,
from human
Expand
View Pricing
M9441
Microsomes from Liver, Pooled,
from mouse(CD-1), male
Expand
View Pricing
M9191
Microsomes from Liver, Pooled,
from rat(Sprague-Dawley), female
Expand
View Pricing
M0567
Microsomes from Liver, Pooled,
from human male
Expand
View Pricing
M9566
Microsomes from Liver, Pooled,
from mouse(CD-1), female
Expand
View Pricing
M0692
Microsomes from Liver, Pooled,
from human female
Expand
View Pricing