Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Micro particles based on silicon dioxide
Synonym(s):
Beads based on silicon dioxide, microsize, Silicon dioxide beads
Products
Brand
Supelco (4)
Physical Form
Format
Quality Segment
Application
Particle Size (µm)
Reference Material Type
Special Grade
analytical standard (4)
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
44054
Micro particles based on silicon dioxide,
size: 5 μm
Expand
View Pricing
56798
Micro particles based on silicon dioxide,
size: 1.0 μm
Expand
View Pricing
56796
Micro particles based on silicon dioxide,
size: 0.5 μm
Expand
View Pricing
81108
Micro particles based on silicon dioxide,
size: 2 μm
Expand
View Pricing