Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Microcapillary tube, calibrated
Products
Product Category
tubes (6)
Brand
Sigma-Aldrich (5)
Color
Manufacturer
Package Size
Filter Products
Showing 1-6 of 6
Compare
Product Number
Description
Pricing
P0549
Microcapillary tube, calibrated,
capacity 1-5 μL
Expand
View Pricing
P0674
Microcapillary tube, calibrated,
capacity 10 μL
Expand
View Pricing
P0799
Microcapillary tube, calibrated,
capacity 20 μL
Expand
View Pricing
P1049
Microcapillary tube, calibrated,
capacity 50 μL
Expand
View Pricing
P1174
Microcapillary tube, calibrated,
capacity 100 μL
Expand
View Pricing
P0924
Microcapillary tube, calibrated,
capacity 40 μL
Expand
View Pricing