Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Mettler Toledo FiveGo
™
pH / mV portable meter
Products
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
MT30266946
Mettler Toledo FiveGo
™
pH / mV portable meter,
model, F2, meter only, AC/DC input 230 V AC, universal plug set
Expand
View Pricing
MT30266889
Mettler Toledo FiveGo
™
pH / mV portable meter,
model, F2, Kit including plastic LE438-IP67 3-in-1 pH electrode, AC/DC input 230 V AC, universal plug set
Expand
View Pricing
MT30266882
Mettler Toledo FiveGo
™
pH / mV portable meter,
model, F2, Field Kit including plastic LE438-IP67 3-in-1 pH electrode and hard-shell carrying case, AC/DC input 230 V AC, universal plug set
Expand
View Pricing
MT30266881
Mettler Toledo FiveGo
™
pH / mV portable meter,
model, F2, Food Kit including LE427-IP67 puncture pH electrode and hard-shell carrying case, AC/DC input 230 V AC, universal plug set
Expand
View Pricing