Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Mettler Toledo Electrolyte for ISE
Products
Manufacturer
Physical Form
Volume (ml)
Filter Products
Showing 1-17 of 17
Compare
Product Number
Description
Pricing
MT51340031
Mettler Toledo Electrolyte for ISE,
For Fluoride ISE, volume 20 mL
Expand
View Pricing
MT51340035
Mettler Toledo Electrolyte for ISE,
For Ammonium ISE, volume 20 mL
Expand
View Pricing
MT51107898
Mettler Toledo Electrolyte for ISE,
For Iodide ISE, volume 20 mL
Expand
View Pricing
MT51107890
Mettler Toledo Electrolyte for ISE,
For Fluoroborate ISE, volume 20 mL
Expand
View Pricing
MT51340032
Mettler Toledo Electrolyte for ISE,
For Calcium ISE, volume 20 mL
Expand
View Pricing
MT51340033
Mettler Toledo Electrolyte for ISE,
For Potassium ISE, volume 20 mL
Expand
View Pricing
MT51340030
Mettler Toledo Electrolyte for ISE,
For Chloride ISE, volume 20 mL
Expand
View Pricing
MT51340034
Mettler Toledo Electrolyte for ISE,
For Nitrate ISE, volume 20 mL
Expand
View Pricing
MT51107892
Mettler Toledo Electrolyte for ISE,
For Barium ISE, volume 20 mL
Expand
View Pricing
MT51107894
Mettler Toledo Electrolyte for ISE,
For Sulfide ISE, volume 20 mL
Expand
View Pricing
MT51107889
Mettler Toledo Electrolyte for ISE,
For Copper ISE, volume 20 mL
Expand
View Pricing
MT51107872
Mettler Toledo Electrolyte for ISE,
For Thiocyanate ISE, volume 20 mL
Expand
View Pricing
MT51340029
Mettler Toledo Electrolyte for ISE,
For Bromide ISE, volume 20 mL
Expand
View Pricing
MT51107893
Mettler Toledo Electrolyte for ISE,
For Cyanide ISE, volume 20 mL
Expand
View Pricing
MT51107891
Mettler Toledo Electrolyte for ISE,
For Cadmium ISE, volume 20 mL
Expand
View Pricing
MT51107881
Mettler Toledo Electrolyte for ISE,
For Lithium ISE, volume 20 mL
Expand
View Pricing
MT51107875
Mettler Toledo Electrolyte for ISE,
For Lead ISE, volume 20 mL
Expand
View Pricing