Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Mettler-Toledo certified buffer
Products
Product Category
buffers (7)
Manufacturer
Physical Form
Volume (ml)
Sterility
Filter Products
Showing 1-7 of 7
Compare
Product Number
Description
Pricing
MT51350032
Mettler-Toledo certified buffer,
pH 4.01, volume 250 mL
Expand
View Pricing
MT51350042
Mettler-Toledo certified buffer,
pH 4.01, volume 6 x 250 mL
Expand
View Pricing
MT51350034
Mettler-Toledo certified buffer,
pH 7, volume 250 mL
Expand
View Pricing
MT51350036
Mettler-Toledo certified buffer,
pH 9.21, volume 250 mL
Expand
View Pricing
MT51350044
Mettler-Toledo certified buffer,
pH 7, volume 6 x 250 mL
Expand
View Pricing
MT51350038
Mettler-Toledo certified buffer,
pH 10, volume 250 mL
Expand
View Pricing
MT51350046
Mettler-Toledo certified buffer,
pH 9.21, volume 6 x 250 mL
Expand
View Pricing