Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Methylcyclohexane
Synonym(s):
Hexahydrotoluene
Linear Formula:
C
6
H
11
CH
3
CAS Number:
108-87-2
Molecular Weight:
98.19
EC Number:
203-624-3
Products
Product Category
solvents (3)
Brand
Sigma-Aldrich (4)
Supelco (1)
Boiling Point (°C)
Formula Weight
Physical Form
Product Line
Purity
Format
Quality Segment
Application
Reference Material Type
Special Grade
Anhydrous (1)
analytical standard (1)
Technique
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
M37889
Methylcyclohexane,
ReagentPlus
®
, 99%
Expand
View Pricing
8.06147
Methylcyclohexane,
for synthesis
Expand
View Pricing
259691
Methylcyclohexane,
spectrophotometric grade, 99%
Expand
View Pricing
300306
Methylcyclohexane,
anhydrous, ≥99%
Expand
View Pricing
66294
Methylcyclohexane,
analytical standard
Expand
View Pricing