Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Methyl-β-cyclodextrin
CAS Number:
128446-36-6
Products
Product Category
carbohydrates (3)
polysaccharides (2)
Brand
Sigma-Aldrich (5)
Biological Source
Color
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Purity
Quality Segment
Sterility
Technique
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
C4555
Methyl-β-cyclodextrin,
powder, BioReagent, suitable for cell culture
Expand
View Pricing
332615
Methyl-β-cyclodextrin,
average M
n
1310
Expand
View Pricing
M7439
Methyl-β-cyclodextrin,
Expand
View Pricing
779776
Methyl-β-cyclodextrin,
Wacker Chemie AG, ≥95.0% cyclodextrin basis (calculated)
Expand
View Pricing
779873
Methyl-β-cyclodextrin,
Wacker Chemie AG, ≥98.0% cyclodextrin basis
Expand
View Pricing