Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Methimazole
Synonym(s):
1-Methyl-2-imidazolethiol, 2-Mercapto-1-methylimidazole, Methimazole
Empirical Formula (Hill Notation):
C
4
H
6
N
2
S
CAS Number:
60-56-0
Molecular Weight:
114.17
EC Number:
200-482-4
Products
Brand
Supelco (3)
Formula Weight
Gene Alias
Manufacturer
Melting Point (°C)
Product Line
Purity
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Package Size
API Family
Special Grade
analytical standard (2)
certified reference material (1)
pharmaceutical primary standard (1)
pharmaceutical secondary standard (1)
Technique
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
1411005
Methimazole,
United States Pharmacopeia (USP) Reference Standard
Expand
View Pricing
M8506
Methimazole,
analytical standard
Expand
View Pricing
PHR3037
Methimazole,
pharmaceutical secondary standard, certified reference material
Expand
View Pricing
46429
Methimazole,
VETRANAL
®
, analytical standard
Expand
View Pricing