Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Methanol-d
4
Synonym(s):
Methyl-d
3
alcohol d, Tetradeuteromethanol, Methanol-D4
Linear Formula:
CD
3
OD
CAS Number:
811-98-3
Molecular Weight:
36.07
EC Number:
212-378-6
Products
Product Category
solvents (10)
stable isotopes (6)
alcohols (1)
Brand
Sigma-Aldrich (13)
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Format
Quality Segment
Isotope
Markush Class
Markush Group
Technique
Filter Products
Showing 1-13 of 13
Compare
Product Number
Description
Pricing
151947
Methanol-d
4
,
≥99.8 atom % D
Expand
View Pricing
441384
Methanol-d
4
,
≥99.8 atom % D
Expand
View Pricing
422878
Methanol-d
4
,
99 atom % D
Expand
View Pricing
343803
Methanol-d
4
,
≥99.8 atom % D, contains 0.03 % (v/v) TMS
Expand
View Pricing
1.06028
Methanol-d
4
,
deuteration degree min. 99.8% for NMR spectroscopy MagniSolv
™
Expand
View Pricing
611646
Methanol-d
4
,
≥99.8 atom % D, contains 0.05 % (v/v) TMS
Expand
View Pricing
444758
Methanol-d
4
,
"100%", 99.96 atom % D
Expand
View Pricing
569534
Methanol-d
4
,
anhydrous, ≥99.8 atom % D
Expand
View Pricing
900596
Methanol-d
4
,
reagent grade, ≥99.5 atom % D, ≥99% (CP)
Expand
View Pricing
535435
Methanol-d
4
,
"100%", ≥99.96 atom % D, contains 0.03 % (v/v) TMS
Expand
View Pricing
1.06025
Methanol-d
4
,
deuteration degree min. 99.95% for NMR spectroscopy MagniSolv
™
Expand
View Pricing
439029
Methanol-d
4
,
≥99.8 atom % D, contains 0.1 % (v/v) TMS
Expand
View Pricing
417653
Methanol-d
4
,
≥99.8 atom % D, contains 1 % (v/v) TMS
Expand
View Pricing